Learn More
Medchemexpress LLC HY-P7218 10ug Medchemexpress, IL-33, Mouse CAS: Purity:>98%
SDP
Catalog No. 501970584
Encompass_Preferred
Medchemexpress, HY-P7218 10ug IL-33, Mouse CAS: Recombinant Mouse Interleukin-33 is a tissue-derived nuclear cytokine from the IL-1 family abundantly expressed in endothelial cells, epithelial cells and fibroblast-like cells, both during homeostasis and inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0584 Supplier Medchemexpress LLC Supplier No. HYP721810UG
Medchemexpress, HY-P7218 10ug IL-33, Mouse CAS: Recombinant Mouse Interleukin-33 is a tissue-derived nuclear cytokine from the IL-1 family abundantly expressed in endothelial cells, epithelial cells and fibroblast-like cells, both during homeostasis and inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.