Learn More
AdipoGen IL-33 mouse rec His
SDP
Catalog No. 502563036
Encompass_Preferred
Protein. Mouse IL-33 aa 109-266 is fused at the C-terminus to a His-tag. Source E. coli. Endotoxin content <1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 150mM NaCl. Binds to mouse and human ST2. Purity >90% SDS-PAGE. Interleukin-33 IL-33 HF-NEV IL-1F11, a member of the IL-1 family of cytokines, is expressed by many cell types following pro-inflammatory stimulation and is thought to be released on cell lysis. The 30kDa human IL33 is converted by CASP1 to a 18kDa protein. IL33 binds to and signals through ST2 IL1R1 and its stimulation recruits MYD88, IRAK, IRAK4, and TRAF6, followed by phosphorylation of ERK1 MAPK3/ERK2 MAPK1, p38 MAPK14, and JNK. The ability of IL-33 to target numerous immune cell types, like Th2-like cells, mast cells, and B1 cells, and to induce cytokine and chemokine production underlines its potential in influencing the outcome of a wide range of diseases, such as arthritis.
Catalog No. 50-256-3036 Supplier AdipoGen Supplier No. AG40A0053C010
Protein. Mouse IL-33 aa 109-266 is fused at the C-terminus to a His-tag. Source E. coli. Endotoxin content <1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 150mM NaCl. Binds to mouse and human ST2. Purity >90% SDS-PAGE. Interleukin-33 IL-33 HF-NEV IL-1F11, a member of the IL-1 family of cytokines, is expressed by many cell types following pro-inflammatory stimulation and is thought to be released on cell lysis. The 30kDa human IL33 is converted by CASP1 to a 18kDa protein. IL33 binds to and signals through ST2 IL1R1 and its stimulation recruits MYD88, IRAK, IRAK4, and TRAF6, followed by phosphorylation of ERK1 MAPK3/ERK2 MAPK1, p38 MAPK14, and JNK. The ability of IL-33 to target numerous immune cell types, like Th2-like cells, mast cells, and B1 cells, and to induce cytokine and chemokine production underlines its potential in influencing the outcome of a wide range of diseases, such as arthritis.