Learn More
AdipoGen IL-33 (oxidation resistant) (human) (rec.) (untagged) (10 µg)
SDP
Supplier:  AdipoGen AG40B0160C010
Encompass_Preferred
Protein. Human IL-33 (aa 112-270) is untagged. Amino acids C208 and C232 have been mutated to serine to protect IL-33 from oxidation. Source: E. coli. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to human ST2. Purity: >95% (SDS-PAGE). IL-33 binds to and signals through ST2 (IL-1R1) and its stimulation recruits MYD88, IRAK, IRAK4 and TRAF6, followed by phosphorylation of ERK1 (MAPK3) / ERK2 (MAPK1), p38 (MAPK14) and JNK. The ability of IL-33 to target numerous immune cell types, like Th2-like cells, mast cells and B1 cells, and to induce cytokine and chemokine production underlines its potential in influencing the outcome of a wide range of diseases, such as arthritis, asthma, atopic allergy & anaphylaxis, cardiovascular disease/atherosclerosis, nervous system diseases and sepsis. IL-33 facilitates Treg expansion in vitro and in vivo. Recently, IL-33 has been involved in adipocyte differentiation.
Catalog No. 50-205-3253