Learn More
Medchemexpress LLC IL-34 Human CHO H 10ug
SDP
Catalog No. 5000182451
Encompass_Preferred
IL-34 protein is an important cytokine that promotes the proliferation survival and differentiation of monocytes and macrophages It plays a key role in innate immune responses and inflammatory processes by promoting the release of pro-inflammatory chemokines IL-34 Protein Human (CHO His) is the recombinant human-derived IL-34 protein expressed by CHO with C-His labeled tag
Catalog No. 50-001-82451 Supplier Medchemexpress LLC Supplier No. HYP7321110UG
IL-34 protein is an important cytokine that promotes the proliferation survival and differentiation of monocytes and macrophages It plays a key role in innate immune responses and inflammatory processes by promoting the release of pro-inflammatory chemokines IL-34 Protein Human (CHO His) is the recombinant human-derived IL-34 protein expressed by CHO with C-His labeled tag