Learn More
Medchemexpress LLC IL-34 Mouse HEK293 100ug
SDP
Catalog No. 5000182344
Encompass_Preferred
IL-34 protein is an important cytokine that promotes the proliferation survival and differentiation of monocytes and macrophages It plays a key role in innate immune responses and inflammatory processes by promoting the release of pro-inflammatory chemokines IL-34 Protein Mouse (HEK293 His) is the recombinant mouse-derived IL-34 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-82344 Supplier Medchemexpress LLC Supplier No. HYP73212100UG
IL-34 protein is an important cytokine that promotes the proliferation survival and differentiation of monocytes and macrophages It plays a key role in innate immune responses and inflammatory processes by promoting the release of pro-inflammatory chemokines IL-34 Protein Mouse (HEK293 His) is the recombinant mouse-derived IL-34 protein expressed by HEK293 with C-His labeled tag