Learn More
Sino Biological Human IL-34 Protein (His Tag) 100ug
Supplier:  Sino Biological 10948H08S100
Encompass_Preferred
A DNA sequence encoding the human IL34 (AAH29804.1) (Met1-Pro242) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6279