Learn More
Medchemexpress LLC IL-35 Human HEK293 100ug
SDP
Catalog No. 5000179704
Encompass_Preferred
IL-35 protein plays a key role in immune regulation forming IL-12 cytokine with IL12B or IL-35 cytokine with EBI3/IL27B IL-12 modulates T cell and natural killer cell responses and induces interferon gamma production IL-35 Protein Human (HEK293 Flag-His) is a recombinant protein dimer complex containing human-derived IL-35 protein expressed by HEK293 with C-His C-Flag labeled tag IL-35 Protein Human (HEK293 Flag-His) has molecular weight of 48 3 kDa
Catalog No. 50-001-79704 Supplier Medchemexpress LLC Supplier No. HYP73880100UG
IL-35 protein plays a key role in immune regulation forming IL-12 cytokine with IL12B or IL-35 cytokine with EBI3/IL27B IL-12 modulates T cell and natural killer cell responses and induces interferon gamma production IL-35 Protein Human (HEK293 Flag-His) is a recombinant protein dimer complex containing human-derived IL-35 protein expressed by HEK293 with C-His C-Flag labeled tag IL-35 Protein Human (HEK293 Flag-His) has molecular weight of 48 3 kDa