Learn More
Medchemexpress LLC Interleukin 35 (IL-35), human recombinant (HEK293, C-hFc) | 97.3% | 2UG
SDP
Catalog No. 5000175096
Encompass_Preferred
Recombinant human interleukin-35 (IL-35) is a heterodimeric cytokine produced in HEK293 cells and supplied as a C-terminal human Fc-tagged, lyophilized powder for research applications. The material is formulated from 0.22 μm-filtered PBS (pH 7.4) and quality-tested by batch certificate of analysis for purity and endotoxin.
Catalog No. 50-001-75096 Supplier Medchemexpress LLC Supplier No. HYP703382UG
Recombinant human interleukin-35 (IL-35) is a heterodimeric cytokine produced in HEK293 cells and supplied as a C-terminal human Fc-tagged, lyophilized powder for research applications. The material is formulated from 0.22 μm-filtered PBS (pH 7.4) and quality-tested by batch certificate of analysis for purity and endotoxin.