Learn More
Medchemexpress LLC IL-35 Human HEK293 50ug
SDP
Supplier:  Medchemexpress LLC HYP7388250UG
Encompass_Preferred
IL-35 protein plays a key role in immune regulation forming IL-12 cytokine with IL12B or IL-35 cytokine with EBI3/IL27B IL-12 modulates T cell and natural killer cell responses and induces interferon gamma production IL-35 Protein Human (HEK293 His hFc-Flag) is a recombinant protein dimer complex containing human-derived IL-35 protein expressed by HEK293 with C-hFc C-Flag C-8 His labeled tag
Catalog No. 50-001-79308