Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-36 beta/IL-1F8 H 50ug
SDP

Supplier:  Medchemexpress LLC HYP7186250UG

Encompass_Preferred

Recombinant human IL-36 beta (IL-1F8) is supplied as a lyophilized, tag-free protein for in vitro research use. It engages the IL-36 receptor to activate NF-κB and MAPK pathways and is suitable for biochemical and cell-based assays investigating pro-inflammatory signaling.

  • Tag-free, lyophilized protein for flexible use.
  • Expressed in E. coli and corresponds to residues M1-E157.
  • High purity (≥95% by reducing SDS-PAGE) and approximate molecular weight of ~18 kDa.
  • Stable when stored at -20°C; reconstitute in ddH2O and aliquot for long-term storage.
  • Suitable for signaling studies, receptor binding assays, and cell stimulation experiments.
  • Reconstitution recommendations include optional carrier proteins or 5% trehalose for stabilization.

Catalog No. 50-001-83661


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.