Learn More
Medchemexpress LLC IL-36 beta/IL-1F8 H 50ug
SDP
Catalog No. 5000181420
Encompass_Preferred
IL-36 beta (IL-1F8) a subform of IL-36 family belongs to IL-1 superfamily IL-36 beta mediates inflammatory response L-36 beta binds to IL-36R and recruits the co-receptor IL-1RacP and thereby activating NF- B and MAPK signaling pathways but the activation requires N-terminal cleavage by neutrophil granule-derived proteases[1][2] IL-36 beta/IL-1F8 Protein Human is a recombinant human IL-36 beta without any tag which is produced in E coli
Catalog No. 50-001-81420 Supplier Medchemexpress LLC Supplier No. HYP7254550UG
IL-36 beta (IL-1F8) a subform of IL-36 family belongs to IL-1 superfamily IL-36 beta mediates inflammatory response L-36 beta binds to IL-36R and recruits the co-receptor IL-1RacP and thereby activating NF- B and MAPK signaling pathways but the activation requires N-terminal cleavage by neutrophil granule-derived proteases[1][2] IL-36 beta/IL-1F8 Protein Human is a recombinant human IL-36 beta without any tag which is produced in E coli