Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-36 beta/IL-1F8 H 5ug
SDP

Catalog No. 5000183384
Encompass_Preferred

Recombinant human IL-36 beta (IL-1F8) is a tag-free, full-length cytokine (153 amino acids) produced in E. coli and supplied as a lyophilized powder for research use. The product has a predicted molecular mass of ~17.7 kDa and is characterized by high purity (>95%) by reducing SDS-PAGE. Store at -20°C for long term; after reconstitution, material is stable short-term at 4°C-freeze aliquots at -20°C or -80°C for extended storage. Intended for in vitro studies of IL-36 signaling and inflammatory pathways.

  • Full-length 153 amino acid isoform for native activity.
  • Tag-free recombinant protein to avoid fusion artifacts.
  • Expressed in E. coli for consistent production.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Lyophilized formulation for long-term storage and ease of shipping.
  • Recommended storage and handling preserve bioactivity (freeze aliquots).

Catalog No. 50-001-83384 Supplier Medchemexpress LLC Supplier No. HYP718625UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human IL-36 beta (IL-1F8) is a tag-free, full-length cytokine (153 amino acids) produced in E. coli and supplied as a lyophilized powder for research use. The product has a predicted molecular mass of ~17.7 kDa and is characterized by high purity (>95%) by reducing SDS-PAGE. Store at -20°C for long term; after reconstitution, material is stable short-term at 4°C-freeze aliquots at -20°C or -80°C for extended storage. Intended for in vitro studies of IL-36 signaling and inflammatory pathways.

  • Full-length 153 amino acid isoform for native activity.
  • Tag-free recombinant protein to avoid fusion artifacts.
  • Expressed in E. coli for consistent production.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Lyophilized formulation for long-term storage and ease of shipping.
  • Recommended storage and handling preserve bioactivity (freeze aliquots).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.