Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-36 gamma/IL-1F9 2ug
SDP

Catalog No. 5000185077
Encompass_Preferred

Recombinant human IL-36 gamma (IL-1F9) is a lyophilized cytokine produced in E. coli for research use. The protein induces IL-8 secretion in A431 epithelial cells (ED50 5-20 ng/mL) and is supplied lyophilized from 20 mM Tris-HCl, 100 mM NaCl, 0.1 mM EDTA, pH 8.0. Recommended storage is -20°C, with aliquots at -80°C for extended storage; reconstitution and carrier protein guidance are provided for stability.

  • Contains the human mature sequence S18-D169.
  • Low endotoxin level (<1 EU/μg) suitable for bioassays.
  • Lyophilized powder for convenient storage and transport.
  • Formulated in Tris-HCl with NaCl and EDTA, pH 8.0.
  • Demonstrated biological activity: ED50 5-20 ng/mL in A431 cells.
  • Reconstitution guidance provided; not recommended below 100 μg/mL and carrier protein advised for long-term stability.

Catalog No. 50-001-85077 Supplier Medchemexpress LLC Supplier No. HYP706702UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human IL-36 gamma (IL-1F9) is a lyophilized cytokine produced in E. coli for research use. The protein induces IL-8 secretion in A431 epithelial cells (ED50 5-20 ng/mL) and is supplied lyophilized from 20 mM Tris-HCl, 100 mM NaCl, 0.1 mM EDTA, pH 8.0. Recommended storage is -20°C, with aliquots at -80°C for extended storage; reconstitution and carrier protein guidance are provided for stability.

  • Contains the human mature sequence S18-D169.
  • Low endotoxin level (<1 EU/μg) suitable for bioassays.
  • Lyophilized powder for convenient storage and transport.
  • Formulated in Tris-HCl with NaCl and EDTA, pH 8.0.
  • Demonstrated biological activity: ED50 5-20 ng/mL in A431 cells.
  • Reconstitution guidance provided; not recommended below 100 μg/mL and carrier protein advised for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.