Learn More
Medchemexpress LLC IL-36 gamma/IL-1F9 2ug
SDP
Supplier:  Medchemexpress LLC HYP725442UG
Encompass_Preferred
IL-36 gamma (IL-1F9) a subform of IL-36 family belongs to IL-1 superfamily IL-36 gamma mediates inflammatory response L-36 beta binds to IL-36R and recruits the co-receptor IL-1RacP and thereby activating NF- B and MAPK signaling pathways but the activation requires N-terminal cleavage by neutrophil granule-derived proteases[1][2] IL-36 gamma/IL-1F9 Protein Mouse is a recombinant mouse IL-36 gamma (G13-S164) without any tag which is produced in E coli
Catalog No. 50-001-82985