Learn More
AdipoGen IL-36beta (aa 5-157) (human) (rec.) (untagged) (10 µg)
SDP
Supplier:  AdipoGen AG40B0117C010
Encompass_Preferred
Protein. Human IL-36beta (aa Arg5-Glu157) (isoform 2) is untagged. Source: E. coli. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS + 0.5% trehalose. Purity: >95% (SDS-PAGE). IL-36alpha (IL-1F6), IL-36beta (IL-1F8) and IL-36gamma (IL-1F9) bind to IL-36R (IL-1Rrp2) and IL-1RAcP, activating similar intracellular signals as IL-1 and are inhibited by IL-36Ra. The expression of IL-36 cytokines has been shown to occur at different sites including the lung and skin and can be derived from diverse cell types including keratinocytes, bronchial epithelium as well as macrophages, monocytes and different T cell subsets. IL-36 family members induce the production of proinflammatory cytokines, including IL-12, IL-1beta, IL-6, TNF-alpha and IL-23 in BMDC and CD4 T cells. These cytokines may represent potential targets for immune-mediated inflammatory conditions or, alternatively, could be used as adjuvants in vaccination.
Catalog No. 50-205-3188