Learn More
AdipoGen IL-36beta (human) Matched Pair Detection Set (1 Set)
SDP
Catalog No. 502053340
Encompass_Preferred
Matched Pair Detection Set. Detects human IL-36beta in cell culture supernatant. Colorimetric assay. Sample Type: Cell Culture Supernatant. Range: 15.6 to 1000 pg/ml. Sensitivity: 10 pg/ml. IL-36alpha (IL-1F6), IL-36beta (IL-1F8) and IL-36gamma (IL-1F9) bind to IL-36R (IL-1Rrp2) and IL-1RAcP, activating similar intracellular signals as IL-1. IL-36Ra inhibits the production of proinflammatory cytokines, including IL-12, IL-1beta, IL-6, TNF-alpha and IL-23 induced by IL-36 in BMDC and CD4 T cells. Skin and dendritic cells are targets of the IL-36 interleukins leading to a Th1 response. These cytokines may represent potential targets for immune-mediated inflammatory conditions or, alternatively, could be used as adjuvants in vaccination.
Catalog No. 50-205-3340 Supplier AdipoGen Supplier No. AG46B0009KI01
Matched Pair Detection Set. Detects human IL-36beta in cell culture supernatant. Colorimetric assay. Sample Type: Cell Culture Supernatant. Range: 15.6 to 1000 pg/ml. Sensitivity: 10 pg/ml. IL-36alpha (IL-1F6), IL-36beta (IL-1F8) and IL-36gamma (IL-1F9) bind to IL-36R (IL-1Rrp2) and IL-1RAcP, activating similar intracellular signals as IL-1. IL-36Ra inhibits the production of proinflammatory cytokines, including IL-12, IL-1beta, IL-6, TNF-alpha and IL-23 induced by IL-36 in BMDC and CD4 T cells. Skin and dendritic cells are targets of the IL-36 interleukins leading to a Th1 response. These cytokines may represent potential targets for immune-mediated inflammatory conditions or, alternatively, could be used as adjuvants in vaccination.