Learn More
AdipoGen IL-36gamma (aa 13-164) (mouse) (rec.) (untagged) (10 µg)
SDP
Supplier:  AdipoGen AG40B0100C010
Encompass_Preferred
Protein. Mouse IL-36gamma (aa Gly13-Ser164) is untagged. Source: E. coli. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS + 0.5% Trehalose. Purity: >95% (SDS-PAGE). IL-36alpha (IL-1F6), IL-36beta (IL-1F8) and IL-36gamma (IL-1F9) bind to IL-36R (IL-1Rrp2) and IL-1RAcP, activating similar intracellular signals as IL-1 and are inhibited by IL-36Ra. IL-36 family members induce the production of proinflammatory cytokines, including IL-12, IL-1beta, IL-6, TNF-alpha and IL-23 in BMDC and CD4 T cells, thus promoting neutrophil influx, dendritic cell (DC) activation, polarization of T helper type 1 (Th1) and IL-17-producing T cells (alphabeta T cells and gammadelta T cells) and keratinocyte proliferation. These cytokines may represent potential targets for immune-mediated inflammatory conditions or, alternatively, could be used as adjuvants in vaccination.
Catalog No. 50-205-3165