Learn More
AdipoGen IL-37 aa 46-218hrecHis
SDP
Catalog No. 502562952
Encompass_Preferred
Protein. Human IL-37 aa Val46-Asp218 is fused at the C-terminus to a His-tag. Source E. coli. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Lyophilized from 0.2µm-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 150mM NaCl. Purity >95% SDS-PAGE. IL-37 IL-1F7 IL-1H4 is an IL-1 family member that is expressed only in certain types of human organs and cells such as heart, thymus, testis, kidney, mononuclear cells PBMCs and dendritic cells. IL-37 is an inhibitor of innate immunity with anti-inflammatory properties. It binds to the interleukin-18 receptor IL-18R and its co-receptor SIGIRR. IL-37 is secreted as a full-length and as a processed form starting from amino acid Val46. It plays an important role as a link between innate and adaptive immunity and acts as an inhibitor of autoimmune diseases and tumor growth. IL-37 also inhibited Lipopolysaccharide LPS-induced immunological reaction and LPS-induced osteoclast formation and bone resorption.
Catalog No. 50-256-2952 Supplier AdipoGen Supplier No. AG40A0190C050
Protein. Human IL-37 aa Val46-Asp218 is fused at the C-terminus to a His-tag. Source E. coli. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Lyophilized from 0.2µm-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 150mM NaCl. Purity >95% SDS-PAGE. IL-37 IL-1F7 IL-1H4 is an IL-1 family member that is expressed only in certain types of human organs and cells such as heart, thymus, testis, kidney, mononuclear cells PBMCs and dendritic cells. IL-37 is an inhibitor of innate immunity with anti-inflammatory properties. It binds to the interleukin-18 receptor IL-18R and its co-receptor SIGIRR. IL-37 is secreted as a full-length and as a processed form starting from amino acid Val46. It plays an important role as a link between innate and adaptive immunity and acts as an inhibitor of autoimmune diseases and tumor growth. IL-37 also inhibited Lipopolysaccharide LPS-induced immunological reaction and LPS-induced osteoclast formation and bone resorption.