Learn More
AdipoGen IL-37 human pAb
SDP
Supplier:  AdipoGen AG25A0111C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human IL-37. Detects a band of ~30kDa by Western blot. Source Rabbit. Applications ELISA, IHC, IP, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. IL-37 IL-1F7 IL-1H4 is an IL-1 family member that is expressed only in certain types of human organs and cells such as heart, thymus, testis, kidney, mononuclear cells PBMCs and dendritic cells. IL-37 is an inhibitor of innate immunity with anti-inflammatory properties. It binds to the interleukin-18 receptor IL-18R and its co-receptor SIGIRR. IL-37 is secreted as a full-length and as a processed form starting from amino acid Val46. It plays an important role as a link between innate and adaptive immunity and acts as an inhibitor of autoimmune diseases and tumor growth. IL-37 also inhibited Lipopolysaccharide LPS-induced immunological reaction and LPS-induced osteoclast formation and bone resorption.
Catalog No. 50-256-3209