Learn More
AdipoGen anti-IL-38, mAb (Nhat-1) (100 µg)
SDP
Supplier:  AdipoGen AG20B0070C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human and mouse IL-38. Isotype: Mouse IgG2bkappa. Clone: Nhat-1. Applications: ELISA, WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. IL-38 (IL-1F10) belongs to the IL-1 family of proteins. IL-38 is expressed in heart, placenta, fetal liver, spleen, thymus and tonsil. The expression in a variety of immune tissues and similarity to IL-1Ra suggest a role of IL-38 as a IL-1 family receptor antagonist. Il-38 inhibits IL-36 signaling by binding to IL-1R6. IL-38 (aa 20-152) is released from apoptotic cells to limit inflammatory macrophage and downstream Th17 activation by blocking IL1RAPL1 (Interleukin-1 receptor accessory protein-like 1, TIGIRR-2) signaling. IL-38 is unregulated during some autoimmune diseases such as Systemic Lupus Erythematosus.
Catalog No. 50-205-2983