Learn More
ACROBiosystems ACROBiosystems Cynomolgus IL-3 R alpha / CD123 Protein, His Tag (SPR verified)
Catalog No. 501956495
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus IL-3 R alpha, His Tag (SPR verified) (ILA-C52H6) is expressed from human 293 cells (HEK293). It contains AA Arg 18 - Arg 302 (Accession # G8F3K3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.7 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Cynomolgus IL-3 R alpha, His Tag (SPR verified) (Cat. No. ILA-C52H6) captured on CM5 Chip via anti-His antibody can bind Human IL-3, Tag Free with an affinity constant of 0.271 μM as determined in SPR assay (Biacore 8k) (QC tested).
Catalog No. 50-195-6495 Supplier ACROBiosystems Supplier No. ILAC52H61MG
Recombinant Protein;1MG;Cynomolgus IL-3 R alpha, His Tag (SPR verified) (ILA-C52H6) is expressed from human 293 cells (HEK293). It contains AA Arg 18 - Arg 302 (Accession # G8F3K3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.7 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Cynomolgus IL-3 R alpha, His Tag (SPR verified) (Cat. No. ILA-C52H6) captured on CM5 Chip via anti-His antibody can bind Human IL-3, Tag Free with an affinity constant of 0.271 μM as determined in SPR assay (Biacore 8k) (QC tested).