Learn More
ACROBiosystems ACROBiosystems Mouse IL-3 R alpha / CD123 Protein, His Tag
Supplier:  ACROBiosystems ILAM52H4100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse IL-3 R alpha, His Tag (ILA-M52H4) is expressed from human 293 cells (HEK293). It contains AA Ser 17- Lys 331 (Accession # P26952-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.4 kDa.
Catalog No. 50-195-6465