Learn More
Medchemexpress LLC IL-4 Canine 50ug
SDP
Supplier:  Medchemexpress LLC HYP7321450UG
Encompass_Preferred
The IL-4 protein actively participates in the B cell activation process acting as a costimulator for DNA synthesis It induces the expression of class II MHC molecules on resting B cells enhances IgE and IgG1 secretion and cell surface expression and modulates CD23 expression on lymphocytes and monocytes IL-4 Protein Canine is the recombinant canine-derived IL-4 protein expressed by E coli with no tag
Catalog No. 50-001-82335