Learn More
Cayman Chemical IL-4 InhIbItr 5mg
SDP
Catalog No. 502949723
Encompass_Preferred
An IL-4 inhibitor; binds to IL-4 (Kd = 1.8 μM); inhibits IL-4 activity in a cell-based reporter assay (EC50 = 1.81 μM); selective for IL-4 over IL-13 (EC50 = 18.2 μM); inhibits IL-4-induced STAT6 phosphorylation in THP-1 monocytes (EC50 = 3.1 μM)
Catalog No. 50-294-9723 Supplier Cayman Chemical Supplier No. 325485 MG
Add to Cart
Add to Cart
An IL-4 inhibitor; binds to IL-4 (Kd = 1.8 μM); inhibits IL-4 activity in a cell-based reporter assay (EC50 = 1.81 μM); selective for IL-4 over IL-13 (EC50 = 18.2 μM); inhibits IL-4-induced STAT6 phosphorylation in THP-1 monocytes (EC50 = 3.1 μM)