Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-4 Mouse HEK293 250ug
SDP

Catalog No. 5000185305
Encompass_Preferred

Recombinant mouse interleukin-4 produced in HEK293 cells with a C-terminal 6xHis tag. Supplied lyophilized in a stabilizing buffer with trehalose, the protein is >95% pure by reducing SDS-PAGE and displays an apparent molecular weight of approximately 19-22 kDa due to glycosylation. Available in multiple sizes including a 250 μg vial; storage and reconstitution guidance is provided for laboratory use.

  • Produced in HEK293 cells to preserve native glycosylation.
  • Contains a C-terminal 6xHis tag for detection and purification.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Apparent molecular weight approximately 19-22 kDa due to glycosylation.
  • Lyophilized with trehalose for improved stability during storage.
  • Recommended reconstitution at ≥100 μg/mL; aliquot and store at -20°C or -80°C with carrier protein for long-term stability.

Catalog No. 50-001-85305 Supplier Medchemexpress LLC Supplier No. HYP70653250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse interleukin-4 produced in HEK293 cells with a C-terminal 6xHis tag. Supplied lyophilized in a stabilizing buffer with trehalose, the protein is >95% pure by reducing SDS-PAGE and displays an apparent molecular weight of approximately 19-22 kDa due to glycosylation. Available in multiple sizes including a 250 μg vial; storage and reconstitution guidance is provided for laboratory use.

  • Produced in HEK293 cells to preserve native glycosylation.
  • Contains a C-terminal 6xHis tag for detection and purification.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Apparent molecular weight approximately 19-22 kDa due to glycosylation.
  • Lyophilized with trehalose for improved stability during storage.
  • Recommended reconstitution at ≥100 μg/mL; aliquot and store at -20°C or -80°C with carrier protein for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.