Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-4 (mouse) | >95.0% | 250UG
SDP

Catalog No. 5000176398
Encompass_Preferred

Recombinant, glycosylated mouse interleukin-4 expressed in HEK293 cells and supplied lyophilized. It promotes proliferation and differentiation of B lymphocytes and regulates multiple immune cell types; characterized for purity and bioactivity for research applications.

  • Glycosylated recombinant protein produced in HEK293 cells.
  • High purity (>95%) by reducing SDS-PAGE.
  • Apparent molecular weight ~19.2 kDa; predicted mass 13.6 kDa.
  • Lyophilized formulation with 8% trehalose for stability.
  • High bioactivity: ED50 0.1049 ng/mL, specific activity 9.533x10^6 units/mg.
  • Store lyophilized at -20°C; aliquot and freeze at -80°C for long-term storage.

Catalog No. 50-001-76398 Supplier Medchemexpress LLC Supplier No. HYP701093250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant, glycosylated mouse interleukin-4 expressed in HEK293 cells and supplied lyophilized. It promotes proliferation and differentiation of B lymphocytes and regulates multiple immune cell types; characterized for purity and bioactivity for research applications.

  • Glycosylated recombinant protein produced in HEK293 cells.
  • High purity (>95%) by reducing SDS-PAGE.
  • Apparent molecular weight ~19.2 kDa; predicted mass 13.6 kDa.
  • Lyophilized formulation with 8% trehalose for stability.
  • High bioactivity: ED50 0.1049 ng/mL, specific activity 9.533x10^6 units/mg.
  • Store lyophilized at -20°C; aliquot and freeze at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.