Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-4 protein, mouse (HEK293) | >95.0% | 50UG
SDP

Catalog No. 5000176401
Encompass_Preferred

IL-4 protein, mouse (HEK293) is a recombinant HEK293-expressed interleukin-4 supplied as a lyophilized, tag-free protein for research use. It promotes B-cell proliferation, differentiation, and cell-surface protein modulation, and is suited for in vitro immune-response and cell-biology assays.

  • Recombinant mouse interleukin-4 expressed in HEK293 cells.
  • Tag-free, lyophilized powder for convenient storage and handling.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Glycosylated; apparent molecular weight ~19.2 kDa by SDS-PAGE.
  • Low endotoxin level (<0.2 EU/μg) suitable for sensitive assays.
  • Reconstitute at ≥100 μg/mL; store aliquots at -20 °C or -80 °C for extended storage.

Catalog No. 50-001-76401 Supplier Medchemexpress LLC Supplier No. HYP70109350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

IL-4 protein, mouse (HEK293) is a recombinant HEK293-expressed interleukin-4 supplied as a lyophilized, tag-free protein for research use. It promotes B-cell proliferation, differentiation, and cell-surface protein modulation, and is suited for in vitro immune-response and cell-biology assays.

  • Recombinant mouse interleukin-4 expressed in HEK293 cells.
  • Tag-free, lyophilized powder for convenient storage and handling.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Glycosylated; apparent molecular weight ~19.2 kDa by SDS-PAGE.
  • Low endotoxin level (<0.2 EU/μg) suitable for sensitive assays.
  • Reconstitute at ≥100 μg/mL; store aliquots at -20 °C or -80 °C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.