Learn More
Enzo Life Sciences IL-4 (mouse), (recombinant) (20µg)
SDP
Catalog No. 502013925
Encompass_Preferred
Alternative name: Interleukin-4. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized. Source: Produced in E.coli. Long Term Storage: -20°C.
Catalog No. 50-201-3925 Supplier Enzo Life Sciences Supplier No. ENZPRT1330020
Alternative name: Interleukin-4. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized. Source: Produced in E.coli. Long Term Storage: -20°C.