Learn More
Genscript Corporation Antibody Protein Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0315010
Encompass
Interleukin-4 Receptor also known as IL-4RA and CD124 is a transmembrane glycoprotein belonging to the class I receptor family. It is highly expressed by activated T-cells. IL-4RA couples with gamma chain to form the type I receptor for IL-4. The extracellular domain of IL-4RA binds to IL-4 and antagonizes its activity. IL-4RA plays an important role in Th2 cell differentiation Ig class switching and alternative macrophage activation. It has also been implicated in allergic inflammation tumor progression and atherogenesis.
Catalog No. 50-252-9780