Learn More
Enzo Life Sciences IL-4 Recombinant monoclonal antibody (11B11) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5480200
Encompass_Preferred
Clone: 11B11. Isotype: IgGκ. Immunogen: BSF-1 / IL-4 purified from serum-free supernatants of cultured EL-4 cells (a C57BL/6 thymoma) induced with 4β-phorbol 12β-myristate 12α-acetate for 48h. Species reactivity: Mouse. Specificity: Recongizes a core functional region of mouse IL-4, which prevents this cytokine from interacting with its cognate receptor. Applications: ELISA, Flow Cytometry, IHC in vivo Assay. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3074