Learn More
ACROBiosystems ACROBiosystems Human IL-4 R alpha / CD124 Protein, Fc Tag
Catalog No. 501957084
Encompass_Preferred
Recombinant Protein;1MG;Human IL-4 R alpha, Fc Tag (ILR-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession # P24394-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.4 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-4, Tag Free (Cat. No. IL4-H4218) at 5 μg/mL (100 μL/well)can bind Human IL-4 R alpha, Fc Tag (Cat. No. ILR-H5253) with a linear range of 1-20 ng/mL (QC tested).;Immobilized Human IL-4 R alpha, Fc Tag (Cat. No. ILR-H5253) at 5 μg/mL (100 μL/well)can bind Biotinylated Human IL-4, Avitag,His Tag (Cat. No. IL4-H82E0) with a linear range of 0.1-2 ng/mL (Routinely tested).
Catalog No. 50-195-7084 Supplier ACROBiosystems Supplier No. ILRH52531MG
Recombinant Protein;1MG;Human IL-4 R alpha, Fc Tag (ILR-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession # P24394-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.4 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-4, Tag Free (Cat. No. IL4-H4218) at 5 μg/mL (100 μL/well)can bind Human IL-4 R alpha, Fc Tag (Cat. No. ILR-H5253) with a linear range of 1-20 ng/mL (QC tested).;Immobilized Human IL-4 R alpha, Fc Tag (Cat. No. ILR-H5253) at 5 μg/mL (100 μL/well)can bind Biotinylated Human IL-4, Avitag,His Tag (Cat. No. IL4-H82E0) with a linear range of 0.1-2 ng/mL (Routinely tested).