Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-4 R alpha / CD124 Protein, Avitag™,His Tag
Supplier:  ACROBiosystems ILRH82E9200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human IL-4 R alpha, Avitag,His Tag (ILR-H82E9) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession # NP_000409.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 26.9 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-4, Tag Free (Cat. No. IL4-H4218) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-4 R alpha, Avitag,His Tag (Cat. No. ILR-H82E9) with a linear range of 0.005-0.078 μg/mL (QC tested).
Catalog No. 50-195-7364