Learn More
ACROBiosystems ACROBiosystems Human IL-5 Protein, His Tag
Supplier:  ACROBiosystems IL5H52H350UG
Encompass_Preferred
Recombinant Protein;50UG;Human IL-5, His Tag (IL5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Ser 134 (Accession # P05113-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 15.0 kDa. The protein migrates as 18-23 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-5, His Tag (Cat. No. IL5-H52H3) at 5 μg/mL (100 μL/well) can bind Human IL-5 R alpha, Fc Tag (Cat. No. ILA-H5259) with a linear range of 0.02-0.313 μg/mL (QC tested).
Catalog No. 50-195-6869