Learn More
Enzo Life Sciences IL-5 (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2390100
Encompass_Preferred
Alternative name: Interleukin-5, B-cell differentiation factor I, Eosinophil differentiaion factor, TFR. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered solution containing 10 mM sodium glycinate, pH 8.5. Source: Produced in E. coli. Contains 116/232 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4043