Learn More
Medchemexpress LLC IL-5 Human HEK293 100ug
SDP
Catalog No. 5000185365
Encompass_Preferred
Recombinant human interleukin-5 (IL-5) expressed in HEK293 cells and provided as a His-tagged, lyophilized protein for in vitro research use. It is intended for bioactivity assays, cell stimulation studies, and other laboratory applications requiring biologically active IL-5.
  • Expressed in HEK293 cells with a C-terminal His tag.
  • Lyophilized formulation from buffered saline or Tris-based buffer for stability.
  • Biological activity demonstrated in cell proliferation assays with ED50 of 0.1-0.5 ng/mL.
  • Low endotoxin level, less than 1 EU per microgram.
  • Manufacturer recommends reconstitution to at least 100 μg/mL for optimal handling.
  • Store lyophilized material at -20 °C; aliquot and freeze at -20 °C or -80 °C after reconstitution for extended storage.
  • Suitable for in vitro research applications such as eosinophil activation and signaling studies.
Catalog No. 50-001-85365 Supplier Medchemexpress LLC Supplier No. HYP70612100UG
Recombinant human interleukin-5 (IL-5) expressed in HEK293 cells and provided as a His-tagged, lyophilized protein for in vitro research use. It is intended for bioactivity assays, cell stimulation studies, and other laboratory applications requiring biologically active IL-5.
  • Expressed in HEK293 cells with a C-terminal His tag.
  • Lyophilized formulation from buffered saline or Tris-based buffer for stability.
  • Biological activity demonstrated in cell proliferation assays with ED50 of 0.1-0.5 ng/mL.
  • Low endotoxin level, less than 1 EU per microgram.
  • Manufacturer recommends reconstitution to at least 100 μg/mL for optimal handling.
  • Store lyophilized material at -20 °C; aliquot and freeze at -20 °C or -80 °C after reconstitution for extended storage.
  • Suitable for in vitro research applications such as eosinophil activation and signaling studies.