Learn More
Enzo Life Sciences IL-5 (mouse), (recombinant) (25µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2400025
Encompass_Preferred
Alternative name: Interleukin-5, B-cell differentiation factor I, Eosinophil differentiaion factor, TFR. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered solution containing 20 mM sodium bicarbonate, pH 9. Source: Produced in E. coli. Contains 113/226 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4044