Learn More
Medchemexpress LLC IL-5 Mouse sf9 Hi 5ug
SDP
Catalog No. 5000182268
Encompass_Preferred
IL-5 protein is expressed by T lymphocytes and NK cells and regulates eosinophils affecting their survival differentiation and chemotaxis In addition IL-5 stimulates immunoglobulin production growth and differentiation of activated and resting B cells IL-5 Protein Mouse (sf9 His) is the recombinant mouse-derived IL-5 protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-82268 Supplier Medchemexpress LLC Supplier No. HYP732195UG
IL-5 protein is expressed by T lymphocytes and NK cells and regulates eosinophils affecting their survival differentiation and chemotaxis In addition IL-5 stimulates immunoglobulin production growth and differentiation of activated and resting B cells IL-5 Protein Mouse (sf9 His) is the recombinant mouse-derived IL-5 protein expressed by Sf9 insect cells with C-His labeled tag