Learn More
Medchemexpress LLC IL-5 Rabbit HEK293 100ug | 100ug
SDP
Catalog No. 5000194140
Encompass_Preferred
The IL5 protein is a cytokine that plays a key role in regulating the growth and differentiation of eosinophils a type of white blood cell involved in allergic reactions and asthma Dysregulation of IL5 protein is associated with a variety of allergic and inflammatory diseases IL-5 Protein Rabbit (HEK293 His) is the recombinant Rabbit-derived IL-5 protein expressed by HEK293 with N-6 His labeled tag
Catalog No. 50-001-94140 Supplier Medchemexpress LLC Supplier No. HYP78608100UG
The IL5 protein is a cytokine that plays a key role in regulating the growth and differentiation of eosinophils a type of white blood cell involved in allergic reactions and asthma Dysregulation of IL5 protein is associated with a variety of allergic and inflammatory diseases IL-5 Protein Rabbit (HEK293 His) is the recombinant Rabbit-derived IL-5 protein expressed by HEK293 with N-6 His labeled tag