Learn More
Medchemexpress LLC IL-5 Rabbit HEK293 10ug | 10ug
SDP
Catalog No. 5000194104
Encompass_Preferred
The IL5 protein is a cytokine that plays a key role in regulating the growth and differentiation of eosinophils a type of white blood cell involved in allergic reactions and asthma Dysregulation of IL5 protein is associated with a variety of allergic and inflammatory diseases IL-5 Protein Rabbit (HEK293 His) is the recombinant Rabbit-derived IL-5 protein expressed by HEK293 with N-6 His labeled tag
Catalog No. 50-001-94104 Supplier Medchemexpress LLC Supplier No. HYP7860810UG
The IL5 protein is a cytokine that plays a key role in regulating the growth and differentiation of eosinophils a type of white blood cell involved in allergic reactions and asthma Dysregulation of IL5 protein is associated with a variety of allergic and inflammatory diseases IL-5 Protein Rabbit (HEK293 His) is the recombinant Rabbit-derived IL-5 protein expressed by HEK293 with N-6 His labeled tag