Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-5RA/CD125 ELISA Kit

Catalog No. EHIL5RAX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHIL5RAX5 5 x 96 Tests
EHIL5RA 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EHIL5RAX5 Supplier Invitrogen™ Supplier No. EHIL5RAX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

The Human Interleukin 5 Receptor Alpha/CD125 (Hu IL-5RA) ELISA quantitates Hu IL-5RA in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-5RA. Principle of the method The Human IL-5RA solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Interleukin 5 (IL-5) is a soluble T cell-derived factor, also known as T cell replacing factor (TRF), that induces B cell and eosinophil growth and differentiation. IL-5 exerts its biological activity through the IL-5 receptor (IL-5R), which is composed of two chains: an alpha chain that binds IL-5 with low affinity and a beta chain that does not bind IL-5, but together with the IL-5 alpha chain, constitutes the high affinity IL-5 receptor. The cytoplasmic domain of both the alpha and beta chains is essential for signal transduction. Specifically, the membrane-proximal proline-rich sequence of the cytoplasmic domain of the IL-5R receptor alpha subunit, IL-5Ralpha, is critical for the IL-5 induced proliferative response, expression of nuclear proto-oncogenes and tyrosine phosphorylation of cellular proteins, such as JAK1 and JAK2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q01344
Assay Range 0.15-40 ng/mL
Assay Sensitivity 150 pg/mL
Description CD125 (IL5RA) ELISA Kit, 960 Assays, Human Species, Colorimetric Detection Method, 3568.0 Gene ID, Cell Culture Supernatants, Plasma, Serum Sample Type, IL5RA Gene Symbols, 150 pg/mL Sensitivity, Used For Protein Assays and Analysis
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, ENA- 78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL- 11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-
Sufficient For 5 x 96 assays
Target Species Human
For Use With (Application) Protein Assays and Analysis, Protein Biology, ELISA, Ready-to-Use Immunoassay
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias CD125; CD125 antigen; CDw125; HSIL5R3; IL-5 receptor alpha chain; IL-5 receptor subunit alpha; Il5r; IL-5R subunit alpha; Il5ra; IL-5RA; IL-5R-alpha; interleukin 5 receptor subunit alpha; interleukin 5 receptor type 3; interleukin 5 receptor, alpha; interleukin-5 receptor alpha chain; interleukin-5 receptor subunit alphaCD125; CD125 antigen; CDw125; HSIL5R3; IL-5 receptor alpha chain; IL-5 receptor subunit alpha; Il5r; IL-5R subunit alpha; Il5ra; IL-5RA; IL-5R-alpha; interleukin 5 receptor subunit alpha; interleukin 5 receptor type 3; interleukin 5 receptor, alpha; interleukin-5 receptor alpha chain; interleukin-5 receptor subunit alpha; CD125; CDw125; HSIL5R3; IL5R
Gene ID (Entrez) 3568
Gene Symbol IL5RA
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases IL-5 receptor subunit alpha, IL-5R subunit alpha, IL-5R-alpha, IL-5RA, CDw125
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target IL5RA
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.