Learn More
Genscript Corporation Antibody Protein Proteins Proteins 1 MG 1/EA
Supplier:  Genscript Corporation Z035901
Encompass
Interleukin-5 Receptor alpha (IL-5RAlpha CD125) is a 60 kDa hematopoietin receptor that plays a dominant role in eosinophil biology. Mature human IL-5 RAlpha consists of a 322 aa extracellular domain (ECD) with a WSxWS motif and a four cysteine motif a 20 aa transmembrane segment and a 58 aa cytoplasmic domain.
Catalog No. 50-252-9597