Learn More
ACROBiosystems IL-6 (Biotinylated) - IL-6 R alpha Inhibitor Screening ELISA Kit, 96 tests (EP-146-96tests)
Catalog No. NC3839792
Encompass_Preferred
This competition ELISA kit is designed to screen inhibitors that block human IL-6 binding to IL-6 receptor alpha. Formatted for 96 tests, the assay uses biotinylated IL-6 and streptavidin-HRP for colorimetric detection, and is suitable for inhibitor screening, assay development, and QC workflows.
  • Competition-ELISA format for inhibitor screening of IL-6/IL-6R interaction.
  • Biotinylated IL-6 and purified human IL-6 R alpha provided for direct competition assays.
  • Includes streptavidin-HRP and substrate for colorimetric detection.
  • Supplies sufficient reagents for 96 tests.
  • Unopened kit stores at 2°C-8°C; opened components follow component storage instructions.
  • Suitable for screening antibodies, biologics, and small-molecule inhibitors.
  • Includes buffers and blocking reagents to streamline assay setup.
Catalog No. NC3839792 Supplier ACROBiosystems Supplier No. EP14696TESTS
This competition ELISA kit is designed to screen inhibitors that block human IL-6 binding to IL-6 receptor alpha. Formatted for 96 tests, the assay uses biotinylated IL-6 and streptavidin-HRP for colorimetric detection, and is suitable for inhibitor screening, assay development, and QC workflows.
  • Competition-ELISA format for inhibitor screening of IL-6/IL-6R interaction.
  • Biotinylated IL-6 and purified human IL-6 R alpha provided for direct competition assays.
  • Includes streptavidin-HRP and substrate for colorimetric detection.
  • Supplies sufficient reagents for 96 tests.
  • Unopened kit stores at 2°C-8°C; opened components follow component storage instructions.
  • Suitable for screening antibodies, biologics, and small-molecule inhibitors.
  • Includes buffers and blocking reagents to streamline assay setup.