Learn More
Medchemexpress LLC HY-P7044A 10ug Medchemexpress, IL-6 (CHO-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 501934290
Encompass_Preferred
Medchemexpress, HY-P7044A 10ug IL-6 (CHO-expressed), Human CAS: Recombinant Human Interleukin-6 (CHO-expressed) is a potent inducer of the synthesis of acute phase proteins in adult human hepatocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4290 Supplier Medchemexpress LLC Supplier No. HYP7044A10UG
Medchemexpress, HY-P7044A 10ug IL-6 (CHO-expressed), Human CAS: Recombinant Human Interleukin-6 (CHO-expressed) is a potent inducer of the synthesis of acute phase proteins in adult human hepatocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.