Learn More
Medchemexpress LLC HY-135233 50mg Medchemexpress, N-Succinimidyl-S-acetylthioacetate CAS:76931-93-6 Purity:>98%
SDP
Catalog No. 501969869
Encompass_Preferred
Medchemexpress, HY-P7103A 10ug IL-6 (HEK293-expressed), Rat CAS: Recombinant Rat Interleukin-6 (HEK293-expressed) is a HEK293 cell derived multifunctional cytokine that can regulate various immune and inflammatory responses. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9869 Supplier Medchemexpress LLC Supplier No. HY13523350MG
Medchemexpress, HY-P7103A 10ug IL-6 (HEK293-expressed), Rat CAS: Recombinant Rat Interleukin-6 (HEK293-expressed) is a HEK293 cell derived multifunctional cytokine that can regulate various immune and inflammatory responses. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.