Learn More
Enzo Life Sciences IL-6 (human), high sensitivity ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1780001
Encompass_Preferred
The IL-6 (human), high sensitivity ELISA kit is a colorimetric immunoassay kit with results in 3 hours. Absorbance is read at 450 nm. Measure up to 40 samples in duplicate per 96 well  kit. Alternative name: Interleukin-6. Assay Time: 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-201-3857