Learn More
Sigma Aldrich Fine Chemicals Biosciences IL 6 human Animal componen
Catalog No. 502905136
Encompass
IL-6 (interleukin 6) signals through the IL-6 receptor system that consists of two chains IL-6RÎ and gp130 (membrane glycoprotein 130). The gene IL-6 is mapped to human chromosome 7p15.3. Murine IL-6 is inactive on human cells while both human and murine are equally active on murine cells. Recombinant human IL-6 is a 20.9kDa protein containing 184 amino acid residues.
Catalog No. 50-290-5136 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP309620UG
IL-6 (interleukin 6) signals through the IL-6 receptor system that consists of two chains IL-6RÎ and gp130 (membrane glycoprotein 130). The gene IL-6 is mapped to human chromosome 7p15.3. Murine IL-6 is inactive on human cells while both human and murine are equally active on murine cells. Recombinant human IL-6 is a 20.9kDa protein containing 184 amino acid residues.