Learn More
Enzo Life Sciences IL-6 (mouse), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900045
Encompass_Preferred
The IL-6 (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Alternative Name: Interleukin-6. Sensitivity: 1.01 pg/ml (range 7.81-1,000 pg/ml). Assay Time: <3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Mouse. Contents: Microtiter plate, Conjugate, Antibody, Assay buffer 13, Wash buffer concentrate, Standard, TMB Substrate, Stop solution
Catalog No. 50-658-116