Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IL-6 Uncoated ELISA Kit

Catalog No. 501128863
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
20 x 96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-8863 10 x 96 Tests
50-112-8696 20 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-112-8863 Supplier Invitrogen™ Supplier No. 88706488
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Mouse Interleukin-6 (IL-6) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-6. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08505
Assay Range 8-500 pg/mL
Assay Sensitivity 8 pg/mL
Content And Storage Capture Antibody: Pre-titrated, purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
Standard: Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer: Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme: Pre-titrated Avidin-HRP
Substrate: 1X TMB Solution
Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards
96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22, -76, -86)
Conjugate HRP
Immunoassay Kit Format Uncoated ELISA Kit
Product Type ELISA
Sample Type Cell Lysates, Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6; IL-21
Gene ID (Entrez) 16193
Gene Symbol Il6
Kit Contents Capture Antibody: Pre-titrated, purified antibody, Detection Antibody: Pre-titrated, biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Enzyme: Pre-titrated Avidin-HRP, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86), Purified antibody, Detection Antibody: Pre-titrated, Biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Enzyme: Pre-titrated Avidin-HRP, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
Label or Dye Biotin
Protein Aliases IL-6, B-cell hybridoma growth factor, Interleukin HP-1
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 100 μL, Plasma 100 μL, Serum 100 μL, Supernatant 100 μL
Storage Requirements 2-8°C
Synonym Interleukin-6
Target IL-6
Test Time 1 hr 45 min
Total Assay Time 24 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.