Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IL-6 Rat HEK293 100ug
SDP

Catalog No. 5000197153
Encompass_Preferred

Recombinant rat interleukin-6 (IL-6) produced in HEK293 cells and supplied lyophilized for research applications. It is suitable for studies of immune signaling, inflammation, B-cell differentiation, and cell-based assays. The protein is formulated from 0.22 μm filtered PBS (pH 7.4), shows >95% purity by reducing SDS-PAGE, and has an apparent molecular weight of ~19-26 kDa. Store frozen and reconstitute at recommended concentrations, adding carrier protein for long-term storage.

  • Recombinant rat interleukin-6 expressed in HEK293 cells.
  • Lyophilized formulation for convenient storage and transport.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~19-26 kDa on reducing SDS-PAGE.
  • Recommended reconstitution at or above 100 μg/mL with aliquot storage at -20°C or -80°C using carrier protein for long term.

Catalog No. 50-001-97153 Supplier Medchemexpress LLC Supplier No. HYP7103A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat interleukin-6 (IL-6) produced in HEK293 cells and supplied lyophilized for research applications. It is suitable for studies of immune signaling, inflammation, B-cell differentiation, and cell-based assays. The protein is formulated from 0.22 μm filtered PBS (pH 7.4), shows >95% purity by reducing SDS-PAGE, and has an apparent molecular weight of ~19-26 kDa. Store frozen and reconstitute at recommended concentrations, adding carrier protein for long-term storage.

  • Recombinant rat interleukin-6 expressed in HEK293 cells.
  • Lyophilized formulation for convenient storage and transport.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~19-26 kDa on reducing SDS-PAGE.
  • Recommended reconstitution at or above 100 μg/mL with aliquot storage at -20°C or -80°C using carrier protein for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.