Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-6 Protein, epitope tag free, ultra sensitivity (primary amine labeling)
Supplier: ACROBiosystems IL6H821825UG
Recombinant Protein;25UG;MABSol Biotinylated Human IL-6, epitope tag free, primary amine labeling (IL6-H8218) is expressed from human HEK293 cells. It contains AA Val 30 - Met 212 (Accession # NP_000591). It is the biotinylated form of ActiveMax Human IL-6, Tag Free (IL6-H4218).;The product does NOT contain any epitope tags. The protein has a calculated MW of 20.8 kDa. The protein migrates as 24-27 kDa on a SDS-PAGE gel under reducing (R) condition due to different glycosylation.;Immobilized Human IL-6 R alpha, Fc Tag (Cat. No. ILR-H5259) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-6, epitope tag free, primary amine labeling (Cat. No. IL6-H8218) with a linear range of 1-20 ng/mL (QC tested).;Biotinylated Human IL-6, epitope tag free, primary amine labeling (Cat. No. IL6-H8218) stimulates proliferation of TF-1 human erythroleukemic cell line. The EC50 for this effect is 0.2532-0.4489 ng/mL (Routinely tested).